2-Butyl-1,3-benzoxazol-4-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Amino-2-(but-1-yl)-1,3-benzoxazole

043180-500MG 043180-1G 043180-5G
101859-538EA 921.54 USD
101859-538 101859-540 101859-542
2-Butyl-1,3-benzoxazol-4-amine ≥95%
2-Butyl-1,3-benzoxazol-4-amine

Hazard: Irritant

Formula: C₁₁H₁₄N₂O
CAS Number: 886361-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR