3-[5-(Trifluoromethyl)-2-pyridinyl]benzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

043177-500MG 043177-1G
101859-522EA 556.92 USD
101859-522 101859-524
3-[5-(Trifluoromethyl)-2-pyridinyl]benzenecarboxylic acid ≥95%
3-[5-(Trifluoromethyl)-2-pyridinyl]benzenecarboxylic acid

Hazard: Irritant

Formula: C₁₃H₈F₃NO₂
MDL Number: MFCD09972206
CAS Number: 946409-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR