Ethyl-1-(tert-butyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

043112-500MG 043112-5G
101859-228EA 375.38 USD
101859-228 101859-232
Ethyl-1-(tert-butyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylate ≥95%
Ethyl-1-(tert-butyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₅F₃N₂O₂
MW: 264.25 g/mol
MDL Number: MFCD04973716
CAS Number: 852691-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR