4-(Dimethylamino)-1,1-diphenyl-3-buten-2-one ≥95%

Supplier: Matrix Scientific

043107-500MG 043107-5G
101859-198EA 278.46 USD
101859-198 101859-202
4-(Dimethylamino)-1,1-diphenyl-3-buten-2-one ≥95%
4-(Dimethylamino)-1,1-diphenyl-3-buten-2-one

Hazard: Irritant

Formula: C₁₈H₁₉NO
MW: 265.36 g/mol
MDL Number: MFCD02089618
CAS Number: 103541-08-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR