3-[Cyclohexyl(methyl)amino]-1,1,1-trifluoro-2-propanol ≥95%
Supplier: Matrix Scientific
043047-500MG
043047-5G
101858-872EA
1109.23
USD
101858-872
101858-876
3-[Cyclohexyl(methyl)amino]-1,1,1-trifluoro-2-propanol ≥95%
3-[Cyclohexyl(methyl)amino]-1,1,1-trifluoro-2-propanol
Hazard: Irritant
Formula:
C₁₀H₁₈F₃NO MW: 225.26 g/mol |
MDL Number:
MFCD03001225 CAS Number: 446276-15-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...