Diethyl-2-(4-chlorophenyl)-1,1-cyclopropanedicarboxylate ≥95%

Supplier: Matrix Scientific

043034-500MG 043034-5G
101858-818EA 556.92 USD
101858-818 101858-822
Diethyl-2-(4-chlorophenyl)-1,1-cyclopropanedicarboxylate ≥95%
Diethyl-2-(4-chlorophenyl)-1,1-cyclopropanedicarboxylate

Hazard: Irritant

Formula: C₁₅H₁₇ClO₄
MW: 296.75 g/mol
MDL Number: MFCD03090782
CAS Number: 74444-83-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR