Methyl-4-(isopentyloxy)benzenecarboxylate ≥95%

Supplier: Matrix Scientific

042872-500MG 042872-5G
101857-974EA 278.46 USD
101857-974 101857-978
Methyl-4-(isopentyloxy)benzenecarboxylate ≥95%
Methyl-4-(isopentyloxy)benzenecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₈O₃
MDL Number: MFCD06659534
CAS Number: 408340-71-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR