3,4,6-Trichloropyridazine ≥95%

Supplier: Matrix Scientific

042816-1G 042816-5G
101857-692EA 581.54 USD
101857-692 101857-694
3,4,6-Trichloropyridazine ≥95%
3,4,6-Trichloropyridazine

Hazard: Irritant-Harmful

Formula: C₄HCl₃N₂
MW: 183.4231 g/mol
Melting Pt: 55…58 °C
MDL Number: MFCD00834957
CAS Number: 6082-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR