Pentafluorophenyl 1-ethanesulfonate ≥95%

Supplier: Matrix Scientific

042807-500MG 042807-1G 042807-5G
101857-638EA 375.38 USD
101857-638 101857-640 101857-642
Pentafluorophenyl 1-ethanesulfonate ≥95%
Pentafluorophenyl 1-ethanesulphonate

Hazard: Irritant

Formula: C₈H₅F₅O₃S
MDL Number: MFCD05975129
CAS Number: 597553-60-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR