Ethyl-6-methyl-2-oxo-4-phenyl-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Methyl-2-oxo-4-phenyl-1,2,3,4-tetrahydropyrimidine-5-carboxylic acid ethyl ester, Ethyl 6-Methyl-2-oxo-4-phenyl-1,2,3,4-tetrahydropyrimidine-5-carboxylate

042759-500MG 042759-1G 042759-5G
101857-382EA 181.54 USD
101857-382 101857-384 101857-386
Ethyl-6-methyl-2-oxo-4-phenyl-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%
Ethyl-6-methyl-2-oxo-4-phenyl-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate

Hazard: Irritant

Formula: C₁₄H₁₆N₂O₃
MW: 260.3 g/mol
MDL Number: MFCD00124778
CAS Number: 5395-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR