1,4-Difluoro-2-methyl-5-nitrobenzene ≥95%

Supplier: Matrix Scientific

Synonyms: 2,5-Difluoro-4-nitrotoluene

042737-1G 042737-5G
101857-258EA 15.38 USD
101857-258 101857-260
1,4-Difluoro-2-methyl-5-nitrobenzene ≥95%
1,4-Difluoro-2-methyl-5-nitrobenzene

Hazard: Irritant

Formula: C₇H₅F₂NO₂
MW: 173.1169 g/mol
MDL Number: MFCD06200905
CAS Number: 141412-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR