2-(Ethylsulfanyl)-1-methyl-1H-imidazole-5-carbaldehyde ≥95%
Supplier: Matrix Scientific
042698-500MG
042698-1G
101857-070EA
556.92
USD
101857-070
101857-072
2-(Ethylsulfanyl)-1-methyl-1H-imidazole-5-carbaldehyde ≥95%
2-(Ethylsulfanyl)-1-methyl-1H-imidazole-5-carbaldehyde
Hazard: Irritant
Formula:
C₇H₁₀N₂OS |
MDL Number:
MFCD06797369 CAS Number: 191411-48-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...