1-(Chloromethyl)-2-[(4-chlorophenyl)sulfanyl]benzene ≥95%
Supplier: Matrix Scientific
042682-500MG
042682-1G
042682-5G
101856-980EA
556.92
USD
101856-980
101856-982
101856-984
1-(Chloromethyl)-2-[(4-chlorophenyl)sulfanyl]benzene ≥95%
1-(Chloromethyl)-2-[(4-chlorophenyl)sulfanyl]benzene
Hazard: Irritant
Formula:
C₁₃H₁₀Cl₂S |
MDL Number:
MFCD04117778 CAS Number: 13459-60-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...