3-Chloro-N-[2-(phenylsulfanyl)ethyl]propanamide ≥90%
Supplier: Matrix Scientific
042680-500MG
042680-1G
101856-970EA
375.38
USD
101856-970
101856-972
3-Chloro-N-[2-(phenylsulfanyl)ethyl]propanamide ≥90%
3-Chloro-N-[2-(phenylsulfanyl)ethyl]propanamide
Hazard: Irritant
Formula:
C₁₁H₁₄ClNOS |
MDL Number:
MFCD12025826 CAS Number: 91131-29-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...