Dimethyl-2-allyl-2-(4-nitrophenyl)malonate ≥95%

Supplier: Matrix Scientific

042674-500MG 042674-1G 042674-5G
101856-934EA 278.46 USD
101856-934 101856-936 101856-938
Dimethyl-2-allyl-2-(4-nitrophenyl)malonate ≥95%
Dimethyl-2-allyl-2-(4-nitrophenyl)malonate

Hazard: Irritant

Formula: C₁₄H₁₅NO₆
MDL Number: MFCD03617782
CAS Number: 404597-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR