5-Bromo-3-fluoro-2-pyridine carboxylic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-3-fluoropicolinic acid, 5-Bromo-3-fluoropyridine-2-carboxylic acid

042497-1G 042497-5G
101856-038EA 84.62 USD
101856-038 101856-040
5-Bromo-3-fluoro-2-pyridine carboxylic acid ≥98%
5-Bromo-3-fluoro-2-pyridine carboxylic acid

Hazard: Irritant

Formula: C₆H₃BrFNO₂
MW: 220 g/mol
Melting Pt: ∼ 170 °C (dec.)
MDL Number: MFCD07437865
CAS Number: 669066-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR