5-Bromo-3-chloropyridine-2-carboxylic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-3-chloropicolinic acid

042495-1G 042495-5G 042495-25G
101856-030EA 23.08 USD
101856-030 101856-032 101856-034
5-Bromo-3-chloropyridine-2-carboxylic acid ≥98%
5-Bromo-3-chloropyridine-2-carboxylic acid

Hazard: Irritant

Formula: C₆H₃BrClNO₂
MW: 236.46 g/mol
MDL Number: MFCD12827552
CAS Number: 1189513-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR