3-Bromo-5-chloropyridine-2-carboxylic acid ≥98%
Supplier: Matrix Scientific
Synonyms:
3-Bromo-5-chloropicolinic acid
042494-1G
042494-5G
101856-026EA
43.08
USD
101856-026
101856-028
3-Bromo-5-chloropyridine-2-carboxylic acid ≥98%
3-Bromo-5-chloropyridine-2-carboxylic acid
Hazard: Irritant
Formula:
C₆H₃BrClNO₂ MW: 236.46 g/mol Boiling Pt: 330 °C Density: 1.917 Flash Pt: 153.4 °C |
MDL Number:
MFCD12827551 CAS Number: 1189513-50-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...