5-Bromo-2-pyridineacetic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 2-(5-Bromopyridin-2-yl)acetic acid

042490-1G 042490-5G
101856-006EA 64.62 USD
101856-006 101856-008
5-Bromo-2-pyridineacetic acid ≥98%
5-Bromo-2-pyridineacetic acid

Hazard: Irritant

Formula: C₇H₆BrNO₂
MW: 216.034 g/mol
Boiling Pt: 120…123 °C
MDL Number: MFCD09999983
CAS Number: 192642-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR