3,5,6-Trifluoro-2-hydroxypyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 3,5,6-Trifluoro-2-pyridinol, 3,5,6-Trifluoro-2-pyridone

042489-1G 042489-5G
101856-002EA 72.31 USD
101856-002 101856-004
3,5,6-Trifluoro-2-hydroxypyridine ≥98%
3,5,6-Trifluoro-2-hydroxypyridine

Hazard: Irritant

Formula: C₅H₂F₃NO
MW: 149.07 g/mol
MDL Number: MFCD04114299
CAS Number: 75777-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR