Fmoc-N-Me-Tyr(tBu)-OH

Supplier: Matrix Scientific

Synonyms: N-α-Fmoc-N-α-methyl-O-tert-butyl-L-tyrosine

042485-1G 042485-5G
101855-988EA 144.62 USD
101855-988 101855-990
Fmoc-N-Me-Tyr(tBu)-OH
Fmoc-N-Me-Tyr(tBu)-OH

Hazard: Irritant

Formula: C₂₉H₃₁NO₅
MW: 473.569 g/mol
MDL Number: MFCD02684471
CAS Number: 133373-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR