Fmoc-N-Me-Thr(tBu)-OH

Supplier: Matrix Scientific

042484-1G 042484-5G
101855-984EA 206.15 USD
101855-984 101855-986
Fmoc-N-Me-Thr(tBu)-OH
Fmoc-N-Me-Thr(tBu)-OH

Hazard: Irritant

Formula: C₂₄H₂₉NO₅
MDL Number: MFCD02094431
CAS Number: 117106-20-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR