Fmoc-Nα-methyl-O-benzyl-L-tyrosine
Supplier: Matrix Scientific
042481-1G
042481-5G
101855-972EA
436.92
USD
101855-972
101855-974
Fmoc-Nα-methyl-O-benzyl-L-tyrosine
Fmoc-Nalpha-methyl-O-benzyl-L-tyrosine
Hazard: Irritant
Formula:
C₃₂H₂₉NO₅ MW: 507.59 g/mol |
MDL Number:
MFCD00270746 CAS Number: 428868-52-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...