Fmoc-Nα-methyl-O-benzyl-L-threonine
Supplier: Matrix Scientific
Synonyms:
Fmoc-N-Me-Thr(Bzl)-OH
042480-1G
042480-5G
101855-968EA
363.08
USD
101855-968
101855-970
Fmoc-Nα-methyl-O-benzyl-L-threonine
Fmoc-Nalpha-methyl-O-benzyl-L-threonine
Hazard: Irritant
Formula:
C₂₇H₂₇NO₅ MW: 445.52 g/mol |
MDL Number:
MFCD00235882 CAS Number: 198561-81-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...