Fmoc-Nα-methyl-O-benzyl-L-serine

Supplier: Matrix Scientific

Synonyms: Fmoc-MeSer(Bzl)-OH

042479-1G 042479-5G
101855-964EA 363.08 USD
101855-964 101855-966
Fmoc-Nα-methyl-O-benzyl-L-serine
Fmoc-Nalpha-methyl-O-benzyl-L-serine

Hazard: Irritant

Formula: C₂₆H₂₅NO₅
MW: 431.49 g/mol
MDL Number: MFCD00235880
CAS Number: 84000-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR