Fmoc-Nα-methyl-O-benzyl-D-tyrosine

Supplier: Matrix Scientific

042478-250MG 042478-1G
101855-960EA 412.31 USD
101855-960 101855-962
Fmoc-Nα-methyl-O-benzyl-D-tyrosine
Fmoc-Nalpha-methyl-O-benzyl-D-tyrosine

Hazard: Irritant

Formula: C₃₂H₂₉NO₅
MW: 507.59 g/mol
MDL Number: MFCD04974266
CAS Number: 1354485-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR