Fmoc-Nα-methyl-N-im-trityl-D-histidine

Supplier: Matrix Scientific

042476-250MG 042476-1G
101855-952EA 633.85 USD
101855-952 101855-954
Fmoc-Nα-methyl-N-im-trityl-D-histidine
Fmoc-Nalpha-methyl-N-im-trityl-D-histidine

Hazard: Irritant

Formula: C₄₁H₃₅N₃O₄
MW: 633.75 g/mol
MDL Number: MFCD04974258
CAS Number: 1217608-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR