Fmoc-N-Me-Lys(boc)-OH

Supplier: Matrix Scientific

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-6-((tert-butoxycarbonyl)amino)hexanoic acid

042475-250MG 042475-1G
101855-948EA 823.08 USD
101855-948 101855-950
Fmoc-N-Me-Lys(boc)-OH
Fmoc-N-Me-Lys(boc)-OH

Hazard: Irritant

Formula: C₂₇H₃₄N₂O₆
MW: 482.577 g/mol
MDL Number: MFCD04973182
CAS Number: 197632-76-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR