Fmoc-Nα-methyl-L-tryptophan

Supplier: Matrix Scientific

Synonyms: Fmoc-N-Me-Trp-OH

042473-100MG 042473-1G
101855-940EA 287.69 USD
101855-940 101855-942
Fmoc-Nα-methyl-L-tryptophan
Fmoc-Nalpha-methyl-L-tryptophan

Hazard: Irritant

Formula: C₂₇H₂₄N₂O₄
MW: 440.499 g/mol
MDL Number: MFCD02684512
CAS Number: 112913-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR