Fmoc-Nα-methyl-L-phenylglycine

Supplier: Matrix Scientific

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-2-phenylacetic acid, N-Fmoc-(S)-a-(methylamino)-benzeneacetic acid, Fmoc-N-Me-Phg-OH

042472-1G 042472-5G
101855-936EA 90.77 USD
101855-936 101855-938
Fmoc-Nα-methyl-L-phenylglycine
Fmoc-Nalpha-methyl-L-phenylglycine

Hazard: Irritant

Formula: C₂₄H₂₁NO₄
MW: 387.44 g/mol
MDL Number: MFCD01861296
CAS Number: 574739-36-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR