Fmoc-N-Me-Phe-OH

Supplier: Matrix Scientific

042471-1G 042471-5G
101855-932EA 150.77 USD
101855-932 101855-934
Fmoc-N-Me-Phe-OH
Fmoc-N-Me-Phe-OH

Hazard: Irritant

Formula: C₂₅H₂₃NO₄
MDL Number: MFCD00151938
CAS Number: 77128-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR