Fmoc-N-methyl-L-norvaline

Supplier: Matrix Scientific

Synonyms: Fmoc-N-Me-Nva-OH, N-Fmoc-N-methyl-L-norvaline, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-norvaline, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)pentanoic acid

042470-1G 042470-5G
101855-928EA 226.15 USD
101855-928 101855-930
Fmoc-N-methyl-L-norvaline
Fmoc-N-methyl-L-norvaline

Hazard: Irritant

Formula: C₂₁H₂₃NO₄
MW: 353.418 g/mol
MDL Number: MFCD01861326
CAS Number: 252049-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR