Fmoc-N-Me-Nle-OH

Supplier: Matrix Scientific

Synonyms: N-Fmoc-N-methyl-L-norleucine

042469-1G 042469-5G
101855-924EA 149.23 USD
101855-924 101855-926
Fmoc-N-Me-Nle-OH
Fmoc-N-Me-Nle-OH

Hazard: Irritant

Formula: C₂₂H₂₅NO₄
MW: 367.445 g/mol
MDL Number: MFCD00235878
CAS Number: 112883-42-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR