Fmoc-N-methyl-L-leucine

Supplier: Matrix Scientific

Synonyms: N-Fmoc-N-methyl-L-leucine, Fmoc-N-Me-Leu-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-leucine

042468-5G 042468-25G
101855-922EA 70.77 USD
101855-922 102551-310
Fmoc-N-methyl-L-leucine
Fmoc-N-methyl-L-leucine

Hazard: Irritant

Formula: C₂₂H₂₅NO₄
MW: 367.445 g/mol
MDL Number: MFCD00151933
CAS Number: 103478-62-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR