Fmoc-N-Me-Glu(OtBu)-OH

Supplier: Matrix Scientific

042467-1G 042467-5G
101855-916EA 661.54 USD
101855-916 101855-918
Fmoc-N-Me-Glu(OtBu)-OH
Fmoc-N-Me-Glu(OtBu)-OH

Hazard: Irritant

Formula: C₂₅H₂₉NO₆
MDL Number: MFCD00237028
CAS Number: 200616-40-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR