Fmoc-N-methyl-D-valine

Supplier: Matrix Scientific

Synonyms: N-Fmoc-N-methyl-D-valine, Fmoc-D-N-Me-Val-OH

042463-1G 042463-5G
101855-900EA 263.08 USD
101855-900 101855-902
Fmoc-N-methyl-D-valine
Fmoc-N-methyl-D-valine

Hazard: Irritant

Formula: C₂₁H₂₃NO₄
MDL Number: MFCD00153396
CAS Number: 103478-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR