N-Fmoc-N-methyl-D-phenylalanine
Supplier: Matrix Scientific
Synonyms:
Fmoc-D-N-Me-Phe-OH, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-3-phenylpropanoic acid
042461-1G
042461-5G
101855-892EA
120
USD
101855-892
101855-894
N-Fmoc-N-methyl-D-phenylalanine
N-Fmoc-N-methyl-D-phenylalanine
Hazard: Irritant
Formula:
C₂₅H₂₃NO₄ MW: 401.462 g/mol |
MDL Number:
MFCD00153392 CAS Number: 138775-05-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...