Fmoc-Nα-methyl-D-glutamic acid-γ-t-butylester
Supplier: Matrix Scientific
Synonyms:
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-5-(tert-butoxy)-5-oxopentanoic acid
042458-1G
042458-5G
101855-884EA
823.08
USD
101855-884
101855-886
Fmoc-Nα-methyl-D-glutamic acid-γ-t-butylester
Fmoc-Nalpha-methyl-D-glutamic acid-gamma-t-butylester
Hazard: Irritant
Formula:
C₂₅H₂₉NO₆ MW: 439.51 g/mol |
MDL Number:
MFCD04974254 CAS Number: 1562442-35-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...