Fmoc-N-Me-D-Ala-OH

Supplier: Matrix Scientific

Synonyms: Fmoc-N-methyl-D-alanine

042457-1G 042457-5G
101855-880EA 130.77 USD
101855-880 101855-882
Fmoc-N-Me-D-Ala-OH
Fmoc-N-Me-D-Ala-OH

Hazard: Irritant

Formula: C₁₉H₁₉NO₄
MW: 325.364 g/mol
MDL Number: MFCD00153385
CAS Number: 138774-92-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR