Fmoc-Nα-methyl-4-chloro-D-phenylalanine

Supplier: Matrix Scientific

042455-250MG 042455-1G
101855-872EA 300 USD
101855-872 101855-874
Fmoc-Nα-methyl-4-chloro-D-phenylalanine
Fmoc-Nalpha-methyl-4-chloro-D-phenylalanine

Hazard: Irritant

Formula: C₂₅H₂₂ClNO₄
MDL Number: MFCD04974252
CAS Number: 1217779-77-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR