Fmoc-n-me-arg(mtr)-oh

Supplier: Matrix Scientific

Synonyms: Fmoc-N-Me-Arg(Mtr)-OH

042454-1G 042454-5G
101855-868EA 823.08 USD
101855-868 101855-870
Fmoc-n-me-arg(mtr)-oh
Fmoc-n-me-arg(mtr)-oh

Hazard: Irritant

Formula: C₃₂H₃₈N₄O₇S
MW: 622.737 g/mol
MDL Number: MFCD02094400
CAS Number: 214750-72-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR