Boc-D-MeTyr(Bzl)-OH

Supplier: Matrix Scientific

Synonyms: (R)-3-(4-(Benzyloxy)phenyl)-2-((tert-butoxycarbonyl)(methyl)amino)propanoic acid, N-Boc-N-methyl-O-benzyl-D-tyrosine, Boc-N-Me-D-Tyr(Bzl)-OH, Boc-D-N-Me-Tyr(Bzl)-OH

042448-1G 042448-5G
101855-844EA 547.69 USD
101855-844 101855-846
Boc-D-MeTyr(Bzl)-OH
Boc-D-MeTyr(Bzl)-OH

Hazard: Irritant

Formula: C₂₂H₂₇NO₅
MW: 385.46 g/mol
MDL Number: MFCD02259475
CAS Number: 138774-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR