Boc-Nα-methyl-N-im-trityl-L-histidine
Supplier: Matrix Scientific
042447-250MG
042447-1G
101855-840EA
412.31
USD
101855-840
101855-842
Boc-Nα-methyl-N-im-trityl-L-histidine
Boc-Nalpha-methyl-N-im-trityl-L-histidine
Hazard: Irritant
Formula:
C₃₁H₃₃N₃O₄ MW: 511.63 g/mol |
MDL Number:
MFCD04974255 CAS Number: 1217610-35-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...