Boc-Nα-methyl-N-im-trityl-D-histidine

Supplier: Matrix Scientific

042446-250MG 042446-1G
101855-836EA 510.77 USD
101855-836 101855-838
Boc-Nα-methyl-N-im-trityl-D-histidine
Boc-Nalpha-methyl-N-im-trityl-D-histidine

Hazard: Irritant

Formula: C₃₁H₃₃N₃O₄
MW: 511.63 g/mol
MDL Number: MFCD04974257
CAS Number: 1217682-00-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR