Boc-Nα-methyl-Ne-2-chlorobenzyl-oxycarbonyl-L-lysine

Supplier: Matrix Scientific

042445-250MG 042445-1G
101855-832EA 386.15 USD
101855-832 101855-834
Boc-Nα-methyl-Ne-2-chlorobenzyl-oxycarbonyl-L-lysine
Boc-Nalpha-methyl-Ne-2-chlorobenzyl-oxycarbonyl-L-lysine

Hazard: Irritant

Formula: C₂₀H₂₉ClN₂O₆
MW: 428.92 g/mol
MDL Number: MFCD04974261
CAS Number: 151012-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR