Boc-N-ME-PHG-OH
Supplier: Matrix Scientific
Synonyms:
N-Boc-N-methyl-L-phenylglycine, Boc-N-Me-Phg-OH
042442-1G
042442-5G
101855-820EA
121.54
USD
101855-820
101855-822
Boc-N-ME-PHG-OH
Boc-N-ME-PHG-OH
Hazard: Irritant
Formula:
C₁₄H₁₉NO₄ MW: 265.309 g/mol |
MDL Number:
MFCD00038799 CAS Number: 30925-11-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...