Boc-N-Me-Phe-OH

Supplier: Matrix Scientific

Synonyms: N-Boc-N-methyl-L-phenylalanine

042441-1G 042441-5G 042441-25G
101855-814EA 36.92 USD
101855-814 101855-816 101855-818
Boc-N-Me-Phe-OH
Boc-N-Me-Phe-OH

Hazard: Irritant

Formula: C₁₅H₂₁NO₄
MW: 279.336 g/mol
MDL Number: MFCD00038754
CAS Number: 37553-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR