Boc-N-ME-NVA-OH

Supplier: Matrix Scientific

Synonyms: Boc-N-methyl-L-norvaline, (S)-2-((tert-Butoxycarbonyl)(methyl)amino)pentanoic acid

042440-1G 042440-5G
101855-810EA 275.38 USD
101855-810 101855-812
Boc-N-ME-NVA-OH
Boc-N-ME-NVA-OH

Hazard: Irritant

Formula: C₁₁H₂₁NO₄
MW: 231.292 g/mol
MDL Number: MFCD02094111
CAS Number: 136092-78-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR