Boc-N-methyl-L-leucine

Supplier: Matrix Scientific

Synonyms: Boc-N-Me-Leu-OH

042438-1G 042438-5G
101855-802EA 84.62 USD
101855-802 101855-804
Boc-N-methyl-L-leucine
Boc-N-methyl-L-leucine

Hazard: Irritant

Formula: C₁₂H₂₃NO₄
MW: 245.319 g/mol
MDL Number: MFCD00038522
CAS Number: 53363-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR