Boc-N-Me-Glu(Obzl)-OH
Supplier: Matrix Scientific
042437-1G
042437-5G
101855-798EA
661.54
USD
101855-798
101855-800
Boc-N-Me-Glu(Obzl)-OH
Boc-N-Me-Glu(Obzl)-OH
Hazard: Irritant
Formula:
C₁₈H₂₅NO₆ MW: 351.399 g/mol |
MDL Number:
MFCD00038756 CAS Number: 200615-91-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...